Learn More
DWK Life Sciences Kimble™ KIMAX™-51 Brand Reusable Serological Pipets with Tooled Opening
Click to view available options
Volume (Metric):
0.2 mL
1 mL
5 mL
10 mL
25 mL
Graduations:
0.01 mL
0.1 mL
Color:
Black
Blue
Orange
Red
White
Unit Size:
Pack of 12
Pack of 6
Description
Tempered tip with permanent brown scale fused into uniform bore tubing without etchin
Order Info
Plugs not included.
Compliance
Calibrated To Deliver/Blow Out. Designed from ASTM™ E 1044.
Specifications
Specifications
| Certifications/Compliance | ASTM E1044 |
| Material | Borosilicate Glass |
| Graduated | Yes |
| Graduations | 0.1 mL |
| Length (English) | 13.77 in. |
| Tolerance | ±0.04 mL |
| Color | Blue |
| Volume (Metric) | 5 mL |
| Length (Metric) | 350 mm |
| Type | Serological Pipet |