missing translation for 'onlineSavingsMsg'
Learn More

KIAA0907 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16863995 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16863995 20 μL 20µL
16853995 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16863995 Supplier Proteintech Supplier No. 254191AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNA-binding protein involved in pre-mRNA splicing. Interacts with the PRP19C/Prp19 complex/NTC/Nineteen complex which is part of the spliceosome. Involved in regulating splice site selection. Binds preferentially RNA with A/C rich sequences and poly-C stretches.

Specifications

Antigen KIAA0907
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KHDC4
Gene Accession No. Q3TCX3, Q7Z7F0
Gene Alias BLOM7, KIAA0907, RP11 336K24.1, UPF0469 protein KIAA0907
Gene Symbols Khdc4
Host Species Rabbit
Immunogen KIAA0907 Fusion Protein Ag22217
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22889, 74200
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.