missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KIAA0907 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17100934 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17100934 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17100934 Supplier Invitrogen™ Supplier No. PA5113069

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KIAA0907 Polyclonal Antibody for Western Blot, IHC (P), ELISA

RNA-binding protein involved in pre-mRNA splicing (PubMed:19641227). Interacts with the PRP19C/Prp19 complex/NTC/Nineteen complex which is part of the spliceosome (PubMed:19641227). Involved in regulating splice site selection (PubMed:19641227). Binds preferentially RNA with A/C rich sequences and poly-C stretches (PubMed:23144703). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0907
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KHDC4
Gene Accession No. Q3TCX3, Q7Z7F0
Gene Alias 2810403A07Rik; A430106P18Rik; AI256352; AI451678; BLOM7; Brings lots of money 7; KH homology domain-containing protein 4; Khdc4; KIAA0907; Pre-mRNA splicing factor protein KHDC4; RIKEN cDNA 2810403A07 gene; SNORA80EHG; UPF0469 protein KIAA0907
Gene Symbols KHDC4
Host Species Rabbit
Immunogen Recombinant Human KH homology domain-containing protein 4 protein (1-241AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22889, 74200
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.