missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHDRBS2 Antibody (7G8C10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
196.00 EUR - 552.00 EUR
Specifications
| Antigen | KHDRBS2 |
|---|---|
| Clone | 7G8C10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18145548
|
Novus Biologicals
NBP2-37254 |
0.1 mL |
552.00 EUR
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666995
|
Novus Biologicals
NBP2-37254-0.025ml |
0.025 mL |
196.00 EUR
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KHDRBS2 Monoclonal specifically detects KHDRBS2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| KHDRBS2 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 202559 | |
| Purified recombinant fragment of human KHDRBS2 (AA: 160-349) expressed in E. Coli. | |
| Primary | |
| 39 kDa |
| 7G8C10 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human, Mouse | |
| hSLM-1, KH domain containing, RNA binding, signal transduction associated 2, KH domain-containing, RNA-binding, signal transduction-associated protein 2, MGC26664, Sam68-like mammalian protein 1, SLM1bA535F17.1, SLM-1FLJ38664 | |
| KHDRBS2 | |
| IgG1 | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title