missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kevetrin hydrochloride, MedChemExpress
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
50mg
Description
Kevetrin hydrochloride is a small molecule and activator of the tumor suppressor protein p53, with potential antineoplastic activity.
Specifications
Specifications
| Chemical Name or Material | Kevetrin hydrochloride |
| Physical Form | Solid |
| CAS | 66592-89-0 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C5H10ClN3S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | 4-Isothioureidobutyronitrile hydrochloride thioureidobutyronitrile hydrochloride thioureido butyronitrile hydrochloride |
| Solubility Information | H2O : 100 mg/mL (556.58 mM; Need ultrasonic) ∣DMSO : ≥ 40 mg/mL (222.63 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction