missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, MedChemExpress

Product Code. 16430778
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
This item is not returnable. View return policy

Product Code. 16430778

Brand: Medchem Express HYB0105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ketoconazole (R-41400) is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor.

Ketoconazole (R-41400) is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor.

Specifications

Chemical Name or Material Ketoconazole
Physical Form Solid
CAS 65277-42-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C26H28Cl2N4O4
For Use With (Application) COVID-19-immunoregulation
Synonym Ketoconazol R 41400
Solubility Information DMSO : 25 mg/mL (47.04 mM; Need ultrasonic)
SMILES CC(N1CCN(C2=CC=C(OC[C@@H]3O[C@@](CN4C=CN=C4)(C5=CC=C(Cl)C=C5Cl)OC3)C=C2)CC1)=O
Molecular Weight (g/mol) 531.43
Formula Weight 531.43
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.