Learn More
DWK Life Sciences™ KECK™ Clip for Spherical Joints, POM, S
Product Code. 15884688 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Dark Blue
Dark Red
Light Blue
Orange
Purple
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15884688 Purple Pack of 100
15894688 Light Blue Pack of 100
15804698 Dark Blue Pack of 100
15971581 Dark Red Pack of 100
15814698 Orange Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15884688 Supplier DWK Life Sciences™ Supplier No. 286340501
KECK™ spherical ground joints ensure a secure connection.
KECK™ spherical ground joint clamps represent a favourable alternative to conventional spherical ground joint clamps made of metal. They are easy to put on and take off. The good spring properties of the POM material prevent scratching of the glass surface. An important contribution to safety in the laboratory. The different colours make the sizes easy to distinguish from each other.
  • Excellent dimensional stability and good spring properties
  • No damage to the glass surface during attachment unlike conventional wire clamps
  • Easy, safe and quick to put on and take off
  • Colour coding per size
  • Cost-effective alternative to conventional spherical ground joint clips made of metal
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100
Material POM
For Use With (Equipment) Securing of Spherical Ground Joints
For Use With (Application) Securing of spherical ground joints
Color Purple