Learn More
DWK Life Sciences™ KECK™ Adapter KA, complete, from glass thread to hose connection plastic
Shop All DWK Life Sciences Products
Click to view available options
Size:
GL 14, diameter 4.5 mm
GL 14, diameter 9.0 mm
RD 14, diameter 4.5 mm
RD 14, diameter 9.0 mm
Unit Size:
Each
Pack of 10
Beschreibung
KECK™ adapters for glass threads are available for sizes GL 14 and RD 14. They protect the thread from damage and ensure a fast, secure and tight connection. The threaded cap is applied directly to the external thread of the connection piece. The hose part can be quickly connected to the threaded cap with a 1/4 turn and loosened again.
- Screw the threaded cap directly onto the external thread of the connection piece
- Quick connection and disconnection of the hose part to the threaded cap with only 1/4 turn
- They protect the thread from damage and ensure a fast, secure and tight connection.
- Available for glass threads sizes GL 14 and RD 14
Spezifikation
Spezifikation
| Quantity | 10 |
| For Use With (Equipment) | Connection of a Hose Piece with a Threaded Cap |
| For Use With (Application) | Connection of a hose piece with a threaded cap |
| Size | GL 14, diameter 9.0 mm |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur