missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDU691, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
KDU691, an imidazopyrazine with potent anti-parasitic activity against blood stage schizonts, gametocytes and liver stages, is a Plasmodium PI4K inhibitor. KDU691 selectively inhibits dihydroartemisinin-pretreated Plasmodium falciparum ring-stage parasites.
Specifications
Specifications
| Chemical Name or Material | KDU691 |
| Physical Form | Solid |
| CAS | 1513879-19-0 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H18ClN5O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 150 mg/mL (357.26 mM; Need ultrasonic and warming) |
| SMILES | ClC1=CC=C(N(C(C2=CN3C(C=N2)=NC=C3C4=CC=C(C(NC)=O)C=C4)=O)C)C=C1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction