missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNK2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17169505 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17169505 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17169505 Supplier Invitrogen™ Supplier No. PA5114332

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KCNK2 Polyclonal Antibody for Flow, IHC (P), Western Blot

TREK-1 is a two-pore-domain background potassium channel expressed throughout the central nervous system. It is opened by polyunsaturated fatty acids and lysophospholipids. As well, it is regulated by various neurotransmitters. It has been shown that alterations in the functioning, regulation, or both of the TREK-1 channel may alter mood. TREK-1 is also activated by volatile anesthetics and has been suggested to be an important target in the action of these drugs. Therefore, this particular K+ channel emerges as a potential innovative target for developing new therapeutic agents for anesthesiology and neurology, such as antidepressants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNK2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene KCNK2
Gene Accession No. O95069, P97438, Q920B6
Gene Alias A430027H14Rik; AI848635; arachidonic acid sensitive tandem pore domain potassium channel; EMBL:AAU06141.1, ECO:0000312; Ensembl:ENSRNOP00000003684, ECO:0000312; hTREK-1c; hTREK-1e; ion transport membrane protein; K2p2.1; K2P2.1 potassium channel; KCNK2; kcnk2 {ECO:0000312; Kcnk2 potassium channel; outward rectifying potassium channel protein TREK-1; outward rectifying potassium channel protein TREK-1 {ECO:0000250; Potassium channel subfamily K member 2; potassium channel subfamily K member 2 {ECO:0000250; potassium channel subfamily k member 2 variant 1; potassium channel subfamily k member 2 variant 2; potassium channel, subfamily K, member 2; potassium channel, two pore domain subfamily K, member 2; potassium inwardly-rectifying channel, subfamily K, member 2; potassium two pore domain channel subfamily K member 2; PubMed:16248991}; PubMed:24196565}; RGD:621448}; rTREK1d; Stretch-activated potassium channel TREK-1; stretch-activated potassium channel TREK-1 {ECO:0000303; tandem-pore-domain potassium channel TREK-1; TPKC1; TREK; trek 1; TREK1; TREK-1; TREK-1 K(+) channel subunit; TREK-1 K(+) channel subunit {ECO:0000250; TWIK-related potassium channel 1; two pore domain potassium channel TREK-1; two pore domain potassium channel TREK-1 {ECO:0000303; Two pore potassium channel TPKC1; two pore potassium channel TPKC1 {ECO:0000250; two-pore potassium channel 1; UniProtKB:O95069}
Gene Symbols KCNK2
Host Species Rabbit
Immunogen Synthetic peptide within Human KCNK2 aa 7-56.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16526, 170899, 3776
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.