missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNG2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15684615
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15684615 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15684615 Supplier Invitrogen™ Supplier No. PA550690

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total KCNG2 protein.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily G. This member is a gamma subunit of the voltage-gated potassium channel. The delayed-rectifier type channels containing this subunit may contribute to cardiac action potential repolarization. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KCNG2
Gene Accession No. Q9UJ96
Gene Alias AA591524; cardiac potassium channel subunit; cardiac potassium channel subunit (Kv6.2); KCNF2; Kcng2; Kv6.2; potassium channel, voltage gated modifier subfamily G, member 2; potassium voltage-gated channel modifier subfamily G member 2; potassium voltage-gated channel subfamily G member 2; potassium voltage-gated channel, subfamily G, member 2; voltage-gated potassium channel subunit Kv6.2
Gene Symbols KCNG2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region derived from internal residues of human potassium voltage-gated channel, subfamily G, member 2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240444, 26251
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.