missing translation for 'onlineSavingsMsg'
Learn More

Kartogenin, MedChemExpress

Product Code. 16457076
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457076

Brand: Medchem Express HY16268

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kartogenin (KGN) is an inducer of differentiation of human mesenchymal stem cells into chondrocytes, with an EC50 of 100 nM. Kartogenin binds filamin A, disrupts its interaction with the transcription factor core-binding factor β subunit (CBFβ), and induces chondrogenesis by regulating the CBFβ-RUNX1 transcriptional program. Kartogenin can be used for the research of osteoarthritis (OA).

Kartogenin (KGN) is an inducer of differentiation of human mesenchymal stem cells into chondrocytes, with an EC50 of 100 nM. Kartogenin binds filamin A, disrupts its interaction with the transcription factor core-binding factor β subunit (CBFβ), and induces chondrogenesis by regulating the CBFβ-RUNX1 transcriptional program. Kartogenin can be used for the research of osteoarthritis (OA).

Specifications

Chemical Name or Material Kartogenin
Physical Form Powder
CAS 4727-31-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H15NO3
Synonym KGN
Solubility Information DMSO : ≥ 42 mg/mL (132.35 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)C1=CC=CC=C1C(NC2=CC=C(C3=CC=CC=C3)C=C2)=O
Molecular Weight (g/mol) 317.34
Formula Weight 317.34
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.