missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Kartogenin

Product Code. 16544434 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 16544434

Brand: Thermo Scientific Chemicals 464660010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4727-31-5
Molecular Formula C20H15NO3
Molecular Weight (g/mol) 317.34
InChI Key SLUINPGXGFUMLL-UHFFFAOYSA-N
IUPAC Name 2-({[1,1'-biphenyl]-4-yl}carbamoyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(=O)NC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point >280.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: slightly soluble in dmso and methanol
Formula Weight 317.34
Chemical Name or Material Kartogenin
compliance-icons compliance-icons
Product Identifier
  • Kartogenin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Kartogenin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.