missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KANSL1L Polyclonal Antibody
GREENER_CHOICE

Product Code. 15779141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15779141 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779141 Supplier Invitrogen™ Supplier No. PA561327

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MLYMESPRTV DEKLKGDTFS QMLGFPTPEP TLNTNFVNLK HFGSPQSSKH YQTVFLMRSN STLNKHNENY KQKKLGEPSC NKLKNILYNG SNIQLSKICL SH Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.

C2orf67, also termed as chromosome 2 open reading frame 67, encodes a 705 amino-acid protein. It exists a variant, which encodes 987 amino-acid protein. The C2orf56 may express in lymphoid /immune and nervous system. The specific function of C2orf67 is still non-known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KANSL1L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KANSL1L
Gene Accession No. A0AUZ9, Q5DTI6
Gene Alias 1110028C15Rik; 1190030G24; AI427598; C2orf67; C430010P07Rik; Kansl1l; KAT8 regulatory NSL complex subunit 1 like; KAT8 regulatory NSL complex subunit 1-like; KAT8 regulatory NSL complex subunit 1-like protein; Kiaa4189; male-specific lethal 1 homolog; mKIAA4189; MSL1v2
Gene Symbols KANSL1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KANSL1L. Recombinant protein control fragment (Product #RP-99646).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151050, 367289, 68691
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.