Learn More
Scharlab KALIUMNITRAT PSS 1KG
Brand:  Scharlab PO02851000
KALIUMNITRAT PSS 1KG
Product Code. 11394264
  • ONLINE EXCLUSIVE PRICE
    51.29 EUR
    44.85 EUR / 1kg
    Save 6.44 EUR
    12% Off
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Potassium nitraten extra pure, Pharmpur, Ph Eur, BP, USP
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
SDS
Documents
Documents
Certificates
Promotions
Promotions