Learn More
K858 (Racemic), MedChemExpress
Product Code. 16487766
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487766 100 mg 100mg
16467988 10 mg 10mg
16433917 50 mg 50mg
16410458 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16487766 Supplier Medchem Express Supplier No. HY19966
MedChemExpress K858 Racemic is an ATP-uncompetitive inhibitor of kinesin Eg5 with an IC50 of 1.3 μM.
K858 Racemic is an ATP-uncompetitive inhibitor of kinesin Eg5 with an IC50 of 1.3 μM.
Specifications
Chemical Name or Material K858 (Racemic)
Physical Form Solid
CAS 72926-24-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H15N3O2S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 100 mg/mL (360.57 mM; Need ultrasonic)
SMILES CC(NC1=NN(C(C)=O)C(C2=CC=CC=C2)(C)S1)=O
Molecular Weight (g/mol) 277.34
Formula Weight 277.34
Percent Purity 99.83%
Grade Research