missing translation for 'onlineSavingsMsg'
Learn More

K145 hydrochloride, MedChemExpress

Product Code. 16496936
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496936

Brand: Medchem Express HY15779A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress K145 hydrochloride is a selective, substrate-competitive and orally active SphK2 inhibitor with an IC50 of 4.3 μM and a Ki of 6.4 μM. K145 hydrochloride is inactive against SphK1 and other protein kinases. K145 hydrochloride induces cell apoptosis and has potently antitumor activity.

K145 hydrochloride is a selective, substrate-competitive and orally active SphK2 inhibitor with an IC50 of 4.3 μM and a Ki of 6.4 μM. K145 hydrochloride is inactive against SphK1 and other protein kinases. K145 hydrochloride induces cell apoptosis and has potently antitumor activity.

Specifications

Chemical Name or Material K145 hydrochloride
CAS 1449240-68-9
Color White
Purity Grade Notes Research
Molecular Formula C18H25ClN2O3S
Quantity 100 mg
Solubility Information H2O : 126.7 mg/mL (329.16 mM; Need ultrasonic and warming) ∣DMSO : 50 mg/mL (129.90 mM; Need ultrasonic)
SMILES O=C(N(CCN)C/1=O)SC1=C/CCC2=CC=C(OCCCC)C=C2.[H]Cl
Molecular Weight (g/mol) 384.92
Formula Weight 384.92
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.