Learn More
Kartell™ K120 Rubber Hoses
Click to view available options
Diameter (Metric) Inner:
5 mm
6 mm
8 mm
Unit Size:
6m
Description
- Includes cloth impression surface
- Suitable for air, water, medium concentration acids and alcaline solutions, abrasive products, oxygenized solvents etc.
Specifications
Specifications
| Diameter (Metric) Inner | 6 mm |
| Material | Rubber |
| Diameter (Metric) Outer | 18 mm |
| Overall Length (Metric) | 6 m |
| For Use With (Application) | Suitable for use with air, water, medium concentration acids, alkaline solutions, abrasive products and oxygenized solvents |
| Min. Operating Temperature (Metric) | -40°C |
| Packaging Quantity | 6 m |
| Max. Operating Temperature (Metric) | 70°C |