Description
- Course-knit aramid fiber gloves for applications that involve both thermal and mechanical risks
- A high level of thermal insulation when in direct contact with warm/hot objects
- Aramid fiber construction provides resistance against cuts
- Cotton liner on the inside increases user comfort
- Ideal for light to medium metalworking, glass handling, and automotive industries
Specifications
Specifications
| Coating Material | Uncoated |
| Color | Yellow |
| Latex Free | Yes |
| Material | Aramid Fiber |
| Product Type | Heat-resistant Gloves |
| Size | 12 |
| Style | Full-hand Mitt |
| Temperature (Metric) Max | 250°C |
| EN388 Standard | 2442D |
| Liner Material | Cotton |
| Show More |