missing translation for 'onlineSavingsMsg'
Learn More

K-756, MedChemExpress

Product Code. 16474086
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16474086

Brand: Medchem Express HYU00422

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress K-756 is a direct and selective tankyrase (TNKS) inhibitor, which inhibits the ADP-ribosylation activity of TNKS1 and TNKS2 with IC50s of 31 and 36 nM, respectively.

K-756 is a direct and selective tankyrase (TNKS) inhibitor, which inhibits the ADP-ribosylation activity of TNKS1 and TNKS2 with IC50s of 31 and 36 nM, respectively.

Specifications

Chemical Name or Material K-756
CAS 130017-40-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H27N5O3
Quantity 25 mg
Solubility Information DMSO : 4.55 mg/mL (10.50 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=CC=C2)CN1CC3CCN(C4=C5C=C(OC)C(OC)=CC5=NC=N4)CC3
Molecular Weight (g/mol) 433.5
Formula Weight 433.5
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.