Learn More
Trajan™ Jumbo Syringe
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
785.00 EUR - 1320.00 EUR
Specifications
Material | Acrylic, Acrylic (Barrel) |
---|---|
For Use With (Application) | For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling |
Product Type | Jumbo Syringe |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10751042
|
Trajan™
009920 |
1000 mL |
800.00 EUR
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10300811
|
Trajan™
009910 |
500 mL |
785.00 EUR
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10045470
|
Trajan™
009930 |
2 mL |
1320.00 EUR
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
- Designed for holding and dispensing large volumes of gas
- Heavy duty acrylic barrels
- Easy access to sample for the addition of standards or removal of subsample via secondary port
- Plunger stem can be unscrewed for ease of transportation and storage
- Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Acrylic, Acrylic (Barrel) | |
For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling | |
Jumbo Syringe |