missing translation for 'onlineSavingsMsg'
Learn More

JR-AB2-011, MedChemExpress

Product Code. p-200102133
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475766

Brand: Medchem Express HY122022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JR-AB2-011 is a selective mTORC2 inhibitor with an IC50 value of 0.36 μM. JR-AB2-011 inhibits mTORC2 activity by blocking Rictor-mTOR association (Ki: 0.19 μM). JR-AB2-011 has anti-glioblastoma multiforme (GBM) properties.

JR-AB2-011 is a selective mTORC2 inhibitor with an IC50 value of 0.36 μM. JR-AB2-011 inhibits mTORC2 activity by blocking Rictor-mTOR association (Ki: 0.19 μM). JR-AB2-011 has anti-glioblastoma multiforme (GBM) properties.

Specifications

Chemical Name or Material JR-AB2-011
CAS 2411853-34-2
Color White
Purity Grade Notes Research
Molecular Formula C17H14Cl2FN3OS
Quantity 100 mg
Solubility Information DMSO : 62.5 mg/mL (156.92 mM; Need ultrasonic)
SMILES ClC1=C(Cl)C=C(NC(N2/C(SC(C)C2)=N/C3=CC=C(F)C=C3)=O)C=C1
Molecular Weight (g/mol) 398.28
Formula Weight 398.28
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.