Learn More
(+)-JQ1 PA, MedChemExpress
Product Code. 16477417
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477417 5 mg 5mg
16463206 25 mg 25mg
16440997 50 mg 50mg
16434708 10 mg 10mg
16415536 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16477417 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY112789
MedChemExpress (+)-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1, with an IC50 of 10.4 nM.
(+)-JQ1 PA is a derivative of the Bromodomain and extra-terminal (BET) inhibitor JQ1, with an IC50 of 10.4 nM.
Specifications
Chemical Name or Material (+)-JQ1 PA
Physical Form Solid
CAS 2115701-93-2
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H20ClN5OS
For Use With (Application) Cancer-programmed cell death
Solubility Information Ethanol : 50 mg/mL (114.17 mM; Need ultrasonic)
SMILES O=C(NCC#C)C[C@H]1C2=NN=C(C)N2C3=C(C(C)=C(C)S3)C(C4=CC=C(Cl)C=C4)=N1
Molecular Weight (g/mol) 437.95
Formula Weight 437.95
Percent Purity 98.87%
Grade Research