missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNJ-47117096 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
JNJ-47117096 hydrochloride is potent and selective MELK inhibitor, with an IC50 of 23 nM, also effectively inhibits Flt3, with an IC50 of 18 nM.
Specifications
Specifications
| Chemical Name or Material | JNJ-47117096 hydrochloride |
| CAS | 1610536-69-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H23ClN4O2 |
| Quantity | 100 mg |
| Synonym | MELK-T1 hydrochloride |
| Solubility Information | DMSO : ≥ 250 mg/mL (626.74 mM) ∣H2O : 3.33 mg/mL (8.35 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(NC1=CC=C2CCNCCC2=C1)C3=CC=C(C4=CNN=C4)C=C3OC.Cl |
| Molecular Weight (g/mol) | 398.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction