missing translation for 'onlineSavingsMsg'
Learn More

JNJ 39758979 TFA Salt, TRC

Product Code. 30405841
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405841 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405841 Supplier Toronto Research Chemicals Supplier No. TRCJ2125005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[(3R)-3-Amino-1-pyrrolidinyl]-6-(1-methylethyl)-2-pyrimidinamine
Molecular Formula C11H19N5 . xC2HF3O2
Quantity 5 mg
Synonym 4-[(3R)-3-Amino-1-pyrrolidinyl]-6-(1-methylethyl)-2-pyrimidinamine;
InChI Formula InChI=1S/C11H19N5.C2HF3O2/c1-7(2)9-5-10(15-11(13)14-9)16-4-3-8(12)6-16;3-2(4,5)1(6)7/h5,7-8H,3-4,6,12H2,1-2H3,(H2,13,14,15);(H,6,7)/t8-;/m1./s1
SMILES NC1=NC(C(C)C)=CC(N2C[C@H](N)CC2)=N1.O=C(C(F)(F)F)O
IUPAC Name (R)-4-(3-aminopyrrolidin-1-yl)-6-isopropylpyrimidin-2-amine 2,2,2-trifluoroacetate
Formula Weight g/mol
Percent Purity >95% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.