missing translation for 'onlineSavingsMsg'
Learn More

JNJ-39758979, MedChemExpress

Product Code. p-200096391
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16466017

Brand: Medchem Express HY101189

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ-39758979 is a selective, orally active, and high-affinity histamine H4 receptor antagonist with Kis of 12.5, 5.3, and 25 nM for human, mouse, and monkey histamine H4 receptor, respectively. JNJ-39758979 functionally antagonizes histamine-induced cAMP inhibition with a pA2 of 7.9 in transfected cells. JNJ-39758979 shows good anti-inflammatory and antipruritic activity.

JNJ-39758979 is a selective, orally active, and high-affinity histamine H4 receptor antagonist with Kis of 12.5, 5.3, and 25 nM for human, mouse, and monkey histamine H4 receptor, respectively. JNJ-39758979 functionally antagonizes histamine-induced cAMP inhibition with a pA2 of 7.9 in transfected cells. JNJ-39758979 shows good anti-inflammatory and antipruritic activity.

Specifications

Chemical Name or Material JNJ-39758979
Physical Form Solid
CAS 1046447-90-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H19N5
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 33.33 mg/mL (150.61 mM; Need ultrasonic)
SMILES NC1=NC(C(C)C)=CC(N2C[C@H](N)CC2)=N1
Molecular Weight (g/mol) 221.3
Formula Weight 221.3
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.