missing translation for 'onlineSavingsMsg'
Learn More

JNJ-38877618, MedChemExpress

Product Code. 16425296
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425296

Brand: Medchem Express HY111050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JNJ-38877618 is a potent, highly selective, orally bioavailable Met kinase inhibitor with IC50s of 2 and 3 nM for wild type and mutant Met, respectively.

JNJ-38877618 is a potent, highly selective, orally bioavailable Met kinase inhibitor with IC50s of 2 and 3 nM for wild type and mutant Met, respectively.

Specifications

Chemical Name or Material JNJ-38877618
Physical Form Solid
CAS 943540-74-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H12F2N6
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 5 mg/mL (13.36 mM; Need ultrasonic)
SMILES FC(C1=CC=C2N=CC=CC2=C1)(F)C3=NN=C4C=CC(C5=CC=NC=C5)=NN43
Molecular Weight (g/mol) 374.35
Formula Weight 374.35
Percent Purity 98.26%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.