missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ JIP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13401377
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13401377 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13401377 Supplier Invitrogen™ Supplier No. PA527224

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (94%), Rat (94%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a regulator of the pancreatic beta-cell function. It is highly similar to JIP-1, a mouse protein known to be a regulator of c-Jun amino-terminal kinase (Mapk8). This protein has been shown to prevent MAPK8 mediated activation of transcription factors, and decrease IL-1 beta and MAP kinase kinase 1 (MEKK1) induced apoptosis in pancreatic beta cells. This protein also functions as a DNA-binding transactivator of the glucose transporter GLUT2. RE1-silencing transcription factor (REST) is reported to repress the expression of this gene in insulin-secreting beta cells. This gene is found to be mutated in a type 2 diabetes family, and thus is thought to be a susceptibility gene for type 2 diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JIP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene MAPK8IP1
Gene Accession No. Q9UQF2, Q9WVI9
Gene Alias C-Jun-amino-terminal kinase-interacting protein 1; Ib1; IB-1; Islet-brain 1; islet-brain-1; JIP1; JIP-1; JIP-1-related protein; JNK MAP kinase scaffold protein 1; JNK-interacting protein 1; JRP; Mapk8ip; Mapk8ip1; mitogen activated protein kinase 8 interacting protein; mitogen activated protein kinase 8 interacting protein 1; mitogen-activated protein kinase 8 interacting protein 1; mitogen-activated protein kinase 8-interacting protein 1; mjip-2a; PRKM8 interacting protein; PRKM8IP; protein kinase, mitogen-activated 8 interacting protein; Skip
Gene Symbols MAPK8IP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human JIP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19099, 9479
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.