missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KleenGuard™ Jackson Safety™ G40 Polyurethane Coated Gloves
Specifications
Specifications
| Color Code | Black |
| Color | Black |
| Material | Polyurethane |
| Product Type | Gloves |
| Size | Small |
| For Use With (Application) | Ideally suited use in precision assembly, electronics assembly, controls handling, packaging, and inspection |
| Quantity | 60Pair |
| Resistant To | Abrasion and tear |
| Series | G40 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?