Learn More
GPE Scientific™ J Young Vacuum/Schlenk Flask with O-Ring Tap and Plain Arm
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Description
- Designed with a side arm for evacuating or filling with gases
- For connection to Schlenk lines
- Round-bottomed shape
- Includes plain arm
- Includes O-ring tap
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Includes | O-Ring Tap and Plain Arm |
| Product Type | Vacuum/Schlenk Flask |