Learn More
GPE Scientific™ J Young In Line Taps, Fine Thread
Product Code. 15804894
Click to view available options
Plug Bore:
6 mm
8 mm
10 mm
15 mm
20 mm
25 mm
Unit Size:
Each
Product Code. 15804894
Brand: GPE Scientific™ SPOR/25
Only null left
Add to basket
Add to basket
GPE Scientific™ J Young In Line Taps, Fine Thread are made in various stopcock sizes and with various side arm diameters.
  • In-line format tap
  • Available in various stopcock sizes
  • Fine thread type
  • Available with different diameter side arms
Specifications
Type J Young in line tap, fine thread type
Plug Bore 25 mm
Diameter (Metric) Side Arm ID: 24 mm, OD: 28