Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum
Click to view available options
Measurement Range:
100 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Description
- Cylindrical, oblong shape
- Gas tight container
- Gauged volume
- Standard collecting tube with valved ends
- Includes centralized septum
Specifications
Specifications
| Type | Gas Sample Tube |
| Measurement Range | 2000 mL |