Learn More
Scharlab JÄRN(III)NITRAT-9-HYDRAT PSS 1KG
Brand:  Scharlab HI03401000
JÄRN(III)NITRAT-9-HYDRAT PSS 1KG
Product Code. 11364194
  • ONLINE EXCLUSIVE PRICE
    160.65 EUR
    87.39 EUR / 1kg
    Save 73.26 EUR
    45% Off
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Iron(III) nitrate nonahydrate, extra pure, Reag. Ph Eur
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .
SDS
Documents
Documents
Certificates
Promotions
Promotions