missing translation for 'onlineSavingsMsg'
Learn More

IU1-47, MedChemExpress

Product Code. 16487757
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487757 5 mg 5mg
16425038 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487757 Supplier Medchem Express Supplier No. HY122243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IU1-47 is a potent and specific USP14 inhibitor with an IC50 of 0.6 μM. IU1-47 inhibits IsoT/USP5 with an IC50 of 20 μM. IU1-47 induces tau elimination in cultured neurons.

IU1-47 is a potent and specific USP14 inhibitor with an IC50 of 0.6 μM. IU1-47 inhibits IsoT/USP5 with an IC50 of 20 μM. IU1-47 induces tau elimination in cultured neurons.

Specifications

Chemical Name or Material IU1-47
CAS 670270-31-2
Color White
Purity Grade Notes Research
Molecular Formula C19H23ClN2O
Quantity 5 mg
Solubility Information DMSO : 16.67 mg/mL (50.39 mM; Need ultrasonic)
SMILES CC1=CC(C(CN2CCCCC2)=O)=C(N1C3=CC=C(C=C3)Cl)C
Molecular Weight (g/mol) 330.85
Formula Weight 330.85
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.