missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Itopride hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
Description
Itopride (HSR803) hydrochloride is a potent dopamine-2 antagonist and an acetylcholine esterase (AChE) inhibitor. Itopride hydrochloride enhances gastric motility through both antidopaminergic and anti-acetylcholinesterasic actions, can be used as a gastrointestinal prokinetic agent. Itopride can be used for researching gastro-esophageal reflux disease (GERD).
Specifications
Specifications
| Chemical Name or Material | Itopride hydrochloride |
| CAS | 122892-31-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H27ClN2O4 |
| Quantity | 100 mg |
| Synonym | HSR803 |
| Solubility Information | DMSO : 100 mg/mL (253.24 mM; Need ultrasonic) ∣H2O : 50 mg/mL (126.62 mM; Need ultrasonic) |
| SMILES | O=C(NCC1=CC=C(OCCN(C)C)C=C1)C2=CC=C(OC)C(OC)=C2.Cl |
| Molecular Weight (g/mol) | 394.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction