missing translation for 'onlineSavingsMsg'
Learn More

Itacitinib, MedChemExpress

Product Code. p-200097654
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497248 10 mg 10mg
16477216 100 mg 100mg
16463267 50 mg 50mg
16461236 200 mg 200mg
16459777 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497248 Supplier Medchem Express Supplier No. HY16997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Itacitinib (INCB039110) is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. Itacitinib shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2; Itacitinib is used in the research of myelofibrosis.

Itacitinib (INCB039110) is an orally active and selective inhibitor of JAK1 with an IC50 of 2 nM for human JAK1. Itacitinib shows >20-fold selectivity for JAK1 over JAK2 and >100-fold over JAK3 and TYK2

Specifications

Chemical Name or Material Itacitinib
CAS 1334298-90-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H23F4N9O
Quantity 10 mg
Synonym INCB039110
Solubility Information DMSO : ≥ 30 mg/mL (54.20 mM)
SMILES N#CCC1(N2N=CC(C3=C4C(NC=C4)=NC=N3)=C2)CN(C5CCN(C(C6=C(F)C(C(F)(F)F)=NC=C6)=O)CC5)C1
Molecular Weight (g/mol) 553.51
Formula Weight 553.51
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.