missing translation for 'onlineSavingsMsg'
Learn More

ISX-9, MedChemExpress

Product Code. p-200098760
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16485816

Brand: Medchem Express HY12323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ISX-9 (Isoxazole 9) is a potent inducer of adult neural stem cell differentiation. ISX-9 activates Ca2+ influx through both voltage-gated Ca2+ channels and NMDA receptors and increases neuroD expression. ISX-9 also induces cardiomyogenic differentiation of Notch-activated epicardium-derived cells (NECs).

ISX-9 (Isoxazole 9) is a potent inducer of adult neural stem cell differentiation. ISX-9 activates Ca2+ influx through both voltage-gated Ca2+ channels and NMDA receptors and increases neuroD expression. ISX-9 also induces cardiomyogenic differentiation of Notch-activated epicardium-derived cells (NECs).

Specifications

Chemical Name or Material ISX-9
Physical Form Solid
CAS 832115-62-5
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H10N2O2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym Isoxazole 9
Solubility Information DMSO : ≥ 100 mg/mL (426.86 mM)
SMILES O=C(C1=NOC(C2=CC=CS2)=C1)NC3CC3
Molecular Weight (g/mol) 234.27
Formula Weight 234.27
Percent Purity 98.03%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.