missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isoxsuprine Hydrochloride, Mikromol™
Description
Isoxsuprine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Isoxsuprine hydrochloride |
| CAS | 579-56-6 |
| Analyte or Component Names | Isoxsuprine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H23 N O3 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Isoxsuprine Hydrochloride, (1RS,2SR)-1-(4-Hydroxyphenyl)-2-[[(1SR)-1-methyl-2-phenoxyethyl]amino]propan-1-ol hydrochloride, Norephedrine, p-hydroxy-N-(1-methyl-2-phenoxyethyl)-, hydrochloride (6CI), 1-(p-Hydroxyphenyl)-2-(1'-methyl-2'-phenoxy)ethylaminopropanol-1 hydrochloride, Dilavase, Divadilan, Duvadilan, Duviculine, Isolait, Isoxsuprin hydrochloride, Navilox, Suprilent, Suprox SR, Tidilan, Vadosilan, Vasodilan, Vasoplex, Vasotran, Benzenemethanol, 4-hydroxy-alpha-[1-[(1-methyl-2-phenoxyethyl)amino]ethyl]-, hydrochloride (1:1), Benzenemethanol, 4-hydroxy-alpha-[1-[(1-methyl-2-phenoxyethyl)amino]ethyl]-, hydrochloride (9CI), Benzyl alcohol, p-hydroxy-alpha-[1-[(1-methyl-2-phenoxyethyl)amino]ethyl]-, hydrochloride (8CI) |
| InChI Formula | InChI=1S/C18H23NO3.ClH/c1-13(12-22-17-6-4-3-5-7-17)19-14(2)18(21)15-8-10-16(20)11-9-15;/h3-11,13-14,18-21H,12H2,1-2H3;1H/t13-,14-,18-;/m0./s1 |
| SMILES | Cl.C[C@@H](COc1ccccc1)N[C@@H](C)[C@H](O)c2ccc(O)cc2 |
| IUPAC Name | 4-[(1R,2S)-1-hydroxy-2-[[(2S)-1-phenoxypropan-2-yl]amino]propyl]phenol;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?