Learn More
Isorhamnetin, MedChemExpress
Product Code. 16454887
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16454887
Brand: Medchem Express HYN0776
MedChemExpress Isorhamnetin is a flavonoid compound extracted from the Chinese herb Hippophae rhamnoides L.. Isorhamnetin suppresses skin cancer through direct inhibition of MEK1 and PI3K.
Isorhamnetin is a flavonoid compound extracted from the Chinese herb Hippophae rhamnoides L.. Isorhamnetin suppresses skin cancer through direct inhibition of MEK1 and PI3K.
Specifications
Chemical Name or Material Isorhamnetin
CAS 480-19-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H12O7
Quantity 50 mg
Synonym 3'-Methylquercetin
Solubility Information DMSO : 100 mg/mL (316.20 mM; Need ultrasonic)
SMILES O=C1C(O)=C(C2=CC=C(O)C(OC)=C2)OC3=CC(O)=CC(O)=C13
Molecular Weight (g/mol) 316.26
Formula Weight 316.26
Percent Purity 98.88%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease