missing translation for 'onlineSavingsMsg'
Learn More

Isoprinosine, TRC

Product Code. 17948124
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17948124 100 mg 100mg
17968224 1 g 1g
17990095 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17948124 Supplier Toronto Research Chemicals Supplier No. TRCI822350100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36703-88-5
Molecular Formula C10 H12 N4 O5 . 3 C9 H9 N O3 . 3 C5 H13 N O
Molecular Weight (g/mol) 1115.23
Synonym Inosine 1-(Dimethylamino)-2-propanol 4-(Acetylamino)benzoate, Aviral, Delimmun, Groprinosin, Imunovir, Imunoviral, Inosine Acedobene Dimepranol, Inosine Pranobex, Inosiplex, Isoprinosin, Isoprinosina, Isoviral, Methisoprinol, Metyzoprynol, Modimmunal, NP 113, NPT 10381, Pranosina, Pranosine, Viruxan;
IUPAC Name 4-acetamidobenzoic acid;9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one;1-(dimethylamino)propan-2-ol
SMILES CC(O)CN(C)C.CC(O)CN(C)C.CC(O)CN(C)C.CC(=O)Nc1ccc(cc1)C(=O)O.CC(=O)Nc2ccc(cc2)C(=O)O.CC(=O)Nc3ccc(cc3)C(=O)O.OC[C@H]4O[C@H]([C@H](O)[C@@H]4O)n5cnc6C(=O)NC=Nc56

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H12N4O5.3C9H9NO3.3C5H13NO/c15-1-4-6(16)7(17)10(19-4)14-3-13-5-8(14)11-2-12-9(5)18;3*1-6(11)10-8-4-2-7(3-5-8)9(12)13;3*1-5(7)4-6(2)3/h2-4,6-7,10,15-17H,1H2,(H,11,12,18);3*2-5H,1H3,(H,10,11)(H,12,13);3*5,7H,4H2,1-3H3/t4-,6-,7-,10-;;;;;;/m1....../s1
Formula Weight 1114.5546
Percent Purity >95%
Chemical Name or Material Inosine Pranobex
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.