Learn More
Isocarboxazid, LoGiCal
Product Code. 30460239
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30460239
Brand: LoGiCal LGCFOR1602.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Isocarboxazid
CAS 59-63-2
Analyte or Component Names Isocarboxazid
Shipping Condition Room Temperature
Molecular Formula C12 H13 N3 O2
Certifications/Compliance ISO 9001
Synonym 3-Isoxazolecarboxylic acid, 5-methyl-, 2-(phenylmethyl)hydrazide3-Isoxazolecarboxylic acid, 5-methyl-, 2-benzylhydrazide (6CI,7CI,8CI), 1-Benzyl-2-(5-methyl-3-isoxazolyl-carbonyl)hydrazine, 5-Methyl-3-isoxazolecarboxylic acid 2-benzylhydrazide, BMIH, Benazide, Enerzer, Isocarboxazid, Isocarboxazide, Isocarboxyzid, Marplan, Marplon, NSC 169893, N'-Benzyl N-methyl-5-isoxazolecarboxylhydrazide-3, Ro 5-0831
InChI Formula InChI=1S/C12H13N3O2/c1-9-7-11(15-17-9)12(16)14-13-8-10-5-3-2-4-6-10/h2-7,13H,8H2,1H3,(H,14,16)
Solution Type Neat
SMILES Cc1onc(c1)C(=O)NNCc2ccccc2
IUPAC Name N'-benzyl-5-methyl-1,2-oxazole-3-carbohydrazide
Industry Type Clinical Testing
Molecular Weight (g/mol) 231.25
Quantity 10 mg
Formula Weight 231.1008 g/mol