missing translation for 'onlineSavingsMsg'
Learn More

Isobutyric Acid, TRC

Product Code. 30389844
Change view
Click to view available options
Quantity:
1 g
10 g
Product Code. Quantity
30389844 10 g
30396359 1 g
2 options
This item is not returnable. View return policy

Product Code. 30389844

Brand: Toronto Research Chemicals TRCI78100510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-31-2
Formula Weight 88.0524
InChI Formula InChI=1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)
IUPAC Name 2-methylpropanoic acid
Molecular Formula C4 H8 O2
Molecular Weight (g/mol) 88.11
SMILES CC(C)C(=O)O
Synonym Propanoic acid, 2-methyl-, Isobutyric acid (8CI), 2-Methylpropanoic acid, 2-Methyl-1-propanoic acid, 2-Methylpropionic acid, 2-Propanecarboxylic acid, Dimethylacetic acid, Isobutanoic acid, Isobutyrates, Isopropylformic acid, NSC 62780, i-Butyric acid, iso-Butyric acid, α-Methylpropanoic acid, α-Methylpropionic acid
Chemical Name or Material Isobutyric acid

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.