missing translation for 'onlineSavingsMsg'
Learn More

Isobutyl p-Toluenesulfonate, TRC

Product Code. 30137672
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137672 100 mg 100mg
30136618 500 mg 500mg
30005070 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137672 Supplier Toronto Research Chemicals Supplier No. TRCI779075100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4873-56-7
InChI Formula InChI=1S/C11H16O3S/c1-9(2)8-14-15(12,13)11-6-4-10(3)5-7-11/h4-7,9H,8H2,1-3H3
IUPAC Name isobutyl 4-methylbenzenesulfonate
Molecular Formula C11 H16 O3 S
Molecular Weight (g/mol) 228.31
Chemical Name or Material Isobutyl p-toluenesulfonate

Specifications

Quantity 100 mg
Synonym p-Toluenesulfonic acid isobutyl ester, 2-methylpropyl 4-methylbenzene-1-sulfonate
SMILES CC(C)COS(=O)(=O)c1ccc(C)cc1
Formula Weight 228.082
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.