Learn More
Brady™ ISO Square Polypropylene Safety Sign 'Fire Alarm Flashing Light'
Click to view available options
Dimensions (L x W):
100 x 100 mm
148 x 148 mm
150 x 150 mm
200 x 200 mm
250 x 250 mm
315 x 315 mm
400 x 400 mm
Color:
Photoluminescent on Red
White on Red
Certifications/Compliance:
Class B
Class C
missing translation for 'unitSize'
Each
Description
- To indicate a device that flashes to indicate that the fire alarm has operated
Specifications
Specifications
| Certifications/Compliance | Class B |
| Alternate Part Number | F/F018/NT/PP-PHOLUMB-200X200-1 |
| Finish | Gloss |
| Material | Polypropylene |
| Quantity | 1 Sign |
| Mounting Type | Surface Mount |
| For Use With (Application) | Facility Identification, Safety Identification |
| Height (English) | 7.87 in. |
| Width (English) | 7.87 in. |
| Temperature Range (Metric) | -40°C to +66°C |
| Show More |