Learn More
Brady™ ISO Safety Sign 'Wear Laboratory Coat,' French
Product Code. 17861725
Change view
Click to view available options
Dimensions (L x W):
50 x 150 mm
105 x 297 mm
150 x 450 mm
200 x 600 mm
210 x 148 mm
297 x 210 mm
420 x 297 mm
Material:
Polyester
Polypropylene
Unit Size:
Each
Product Code. Dimensions (L x W) Material unitSize
17861725 210 x 148 mm Polyester Each
17871715 50 x 150 mm Polyester Each
17841725 105 x 297 mm Polyester Each
17851725 150 x 450 mm Polyester Each
17831705 200 x 600 mm Polyester Each
17841775 297 x 210 mm Polyester Each
17891725 420 x 297 mm Polyester Each
17801715 50 x 150 mm Polypropylene Each
17851755 105 x 297 mm Polypropylene Each
17841755 150 x 450 mm Polypropylene Each
17891785 200 x 600 mm Polypropylene Each
17801725 210 x 148 mm Polypropylene Each
17871755 297 x 210 mm Polypropylene Each
17891755 420 x 297 mm Polypropylene Each
14 options
Product Code. 17861725
Brand: Brady™ 311391
Only null left
Add to basket
Add to basket
Safety Sign to signify that a laboratory coat must be worn.
  • Provide visual warning that laboratory coat must be worn in the area
Specifications
Alternate Part Number M/M059/FR797/PE-148X210-1
Finish Gloss
Material Polyester
Quantity 1 Sign
Size (English) 8.27 x 5.83 in.
Adhesive Type Permanent
Mounting Type Surface Mount
For Use With (Application) Facility Identification, Safety Identification
Height (English) 8.27 in.
Thickness 0.099 mm
Width (English) 5.83 in.
Temperature Range (Metric) -40°C to +110°C
Shape Rectangular
Temperature Range (English) -40°F to +230°F
Languages French
Dimensions (L x W) 210 x 148 mm
Displayed Text Wear Laboratory Coat
Product Type Safety Sign
Color White on Blue