missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Irosustat, MedChemExpress
Description
Irosustat is a potent steroid sulfatase inhibitor, with an IC50 of 8 nM, and exhibits anti-breast cancer activity.
Specifications
Specifications
| Chemical Name or Material | Irosustat |
| CAS | 288628-05-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H15NO5S |
| Quantity | 100 mg |
| Synonym | STX64 BN83495 667-Coumate |
| Solubility Information | DMSO : 100 mg/mL (323.27 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | O=C1OC2=CC(OS(=O)(N)=O)=CC=C2C3=C1CCCCC3 |
| Molecular Weight (g/mol) | 309.34 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?