Learn More
Bochem™ Iron Wire Brush
Product Code. p-8005448
Click to view available options
Length (Metric):
260 mm
480 mm
550 mm
1000 mm
Diameter (Metric) Bristle:
30 mm
63 mm
80 mm
For Use With (Application):
For flask
For test tubes
For tubes
Unit Size:
Each
Product Code. 10555221
Brand: Bochem™ 12300
Only null left
Add to basket
Add to basket
Made with zincked iron wire. Bochem™ Iron Wire Brush can be used with flasks and test tubes.
  • Iron wire, zincked
  • Natural bristles
  • Used with flasks and test tubes
  • For cleaning flasks and bottles
Specifications
Product Type Brush
Diameter (Metric) Bristle 30 mm
For Use With (Application) For tubes
Length (Metric) 1000 mm
Length (Metric) Brush 150 mm
Material Iron Wire
Quantity 1 Ea.