Learn More
Usbeck Laborgeraete™ Iron Crucible
Click to view available options
Diameter (Metric) Outer:
50 mm
Unit Size:
Each
Specifications
Specifications
| Material | Iron |
| Diameter (Metric) Outer | 50 mm |
| Height (Metric) | 45mm |
| Product Type | Crucible |