missing translation for 'onlineSavingsMsg'
Learn More

IRL-2500, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18716836
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18716836

Brand: Tocris Bioscience 1838/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent ETB antagonist

Potent endothelin receptor antagonist; shows some selectivity for ETB receptors (IC50 values are 1.3 and 94 nM for ETB and ETA receptors respectively). Inhibits ETB receptor-mediated blood pressure increase and renal vascular resistance in rats in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 169545-27-1
Molecular Formula C36H35N3O4
Synonym chembl72410, 2s-2-2r-2-3,5-dimethylbenzoyl-methylamino-3-4-phenylphenyl propanoyl amino-3-1h-indol-3-yl propanoic acid, tocris-1838, d01ede, n-3-1,1'-biphenyl-4-yl-n-3,5-dimethylbenzoyl-n-methyl-d-alanyl-l-tryptophan, 2s-2-2r-3-1,1'-biphenyl-4-yl-2-1-3,5-dimethylphenyl-n-methylformamido propanamido-3-1h-indol-3-yl propanoic acid, s-2-r-3-biphenyl-4-yl-2-3,5-dimethyl-benzoyl-methyl-amino-propionylamino-3-1h-indol-3-yl-propionic acid
InChI Key UZDORQWMYRRLQV-JHOUSYSJSA-N
SMILES CC1=CC(=CC(=C1)C(=O)N(C)C(CC2=CC=C(C=C2)C3=CC=CC=C3)C(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)O)C
IUPAC Name (2S)-2-[[(2R)-2-[(3,5-dimethylbenzoyl)-methylamino]-3-(4-phenylphenyl)propanoyl]amino]-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 573.693
Quantity 10 mg
PubChem CID 5311192
Formula Weight 573.69
Percent Purity >99%
Chemical Name or Material IRL-2500
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.