Learn More
Irinotecan hydrochloride, MedChemExpress
Product Code. 16431226
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Product Code. 16431226
Brand: Medchem Express HY16562A
MedChemExpress Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.
Irinotecan hydrochloride ((+)-Irinotecan hydrochloride) is a topoisomerase I inhibitor mainly used to treat colon cancer and rectal cancer.
Specifications
Chemical Name or Material Irinotecan hydrochloride
CAS 100286-90-6
Color White
Purity Grade Notes Research
Molecular Formula C33H39ClN4O6
Quantity 200 mg
Synonym (+)-Irinotecan hydrochlorideCPT-11 hydrochloride
Solubility Information DMSO : 125 mg/mL (200.60 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (5.34 mM; Need ultrasonic)
SMILES O=C(N1CCC(N2CCCCC2)CC1)OC3=CC=C4N=C5C(CN6C(C(COC([C@@]7(CC)O)=O)=C7C=C65)=O)=C(CC)C4=C3.[H]Cl
Molecular Weight (g/mol) 623.14
Formula Weight 623.14
Percent Purity 98.03%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death